Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589758 Supplier Invitrogen™ Supplier No. PA589758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPGANYRAGA GAGAGARRPR GARDREEDGG GLEPAAVARD LLRGTSNMSF EELLELQSQV GTKTYKQLVA GNSPKKQASR PPIQNACVAD KHRPLEMSAK IRVPFLRQVV PISKKVARDP RFDDLSGEYN PEVFDKTYQF LNDIRAKEKE LVKKQLKKHL SGEEHEKLQQ LLQRMEQQEM AQQERKQQQE LHLALKQERR AQAQQGHRPY FLKKSEQRQL ALAEKFKELK RSKKLENFLS RKRRRNAGKD RRHLPLSKE; Positive Samples: MCF7, 293T, BxPC-3, HeLa; Cellular Location: Nucleus, nucleolus.

RRP36 functions at an early stage in the processing of 35S preribosomal RNA into the mature 18S species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP36
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RRP36
Gene Accession No. Q96EU6
Gene Alias C6orf153; cf153; dJ20C7.4; HSPC253; ribosomal RNA processing 36; ribosomal RNA processing 36 homolog; ribosomal RNA processing 36 homolog (S. cerevisiae); ribosomal RNA processing protein 36 homolog; RNA processing factor; RRP36
Gene Symbols RRP36
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RRP36 (NP_1491031).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88745
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.