Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597232 Supplier Invitrogen™ Supplier No. PA597232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVARRRKCAA RDPEDRIPSP LGYAAIPIKF SEKQQASHYL YVRAHGVRQG TKSTWPQKRT LFVLNVPPYC TEESLSRLLS TCGLVQSVEL QEKPDLAESP KESRSKFFHP KPVPGFQVAY VVFQKPSGVS AALALKGPLL VSTESHPVKS GIHKWISDYA DSVPDPEALR VEVDTFMEAY DQKIAEEEAK AKEEEGVPDE EGWVKVTRRG RRPVLPRTEA ASLRVLERER RKRSRKELLN FYAWQHRESK MEHLAQLRKK FEEDKQRIEL LRAQRKFRPY; Positive Samples: Mouse heart, Rat heart.

RRP7A (Ribosomal RNA Processing 7 Homolog A) is a Protein Coding gene. Diseases associated with RRP7A include Autosomal Dominant Non-Syndromic Intellectual Disability 1 and Autosomal Dominant Non-Syndromic Intellectual Disability. Among its related pathways are rRNA processing in the nucleus and cytosol and Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleotide binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP7A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RRP7A
Gene Accession No. Q9D1C9
Gene Alias 1110014J01Rik; AA408146; BK126B4.3; CGI-96; CTA-126B4.5; gastric cancer antigen Zg14; Gastric cancer antigen Zg14 homolog; hypothetical protein LOC550241; Kheg1; kidney highly expressed protein 1; RGD1311547; ribosomal RNA processing 7 homolog; ribosomal RNA processing 7 homolog A; ribosomal RNA processing 7 homolog A (S. cerevisiae); ribosomal RNA-processing protein 7 homolog A; RRP7; RRP7A; zgc:110084
Gene Symbols RRP7A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human RRP7A (NP_0565182).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362967, 74778
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.