Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRP8 Mouse anti-Mouse, Human, Unconjugated, Clone: 5C7.1, MilliporeSigma™

Catalog No. MABE1013100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1013-100 100 μg
MABE1013-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1013-100 Supplier MilliporeSigma Supplier No. MABE1013100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RRP8, clone 5C7.1, Cat. No. MABE1013, is a highly specific mouse monoclonal antibody, that targets Ribosomal RNA-processing protein 8 and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP8
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5C7.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ribosomal RNA-processing protein 8;Cerebral protein 1;Nucleomethylin
Gene Symbols RRP8;KIAA0409;NML;hucep-1
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 98 amino acids from the C-terminal half of human Ribosomal RNA-processing protein 8.
Molecular Weight of Antigen ∽53 kDa observed; 50.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_056139
Test Specificity Clone 5C7.1 specifically detects Ribosomal RNA-processing protein 8. It targets an epitope with in the C-terminal half.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.