Learn More
RRP8 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304202A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Cerebral 1 also called hucep-1, KIAA0409 or RRP8, is an essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD+/NADP+ ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at ′Lys-9′ (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.Specifications
RRP8 | |
Polyclonal | |
Unconjugated | |
RRP8 | |
Ribosomal RNA-processing protein 8, Cerebral protein 1, Nucleomethylin, RRP8, KIAA0409, NML, hucep-1, HUCE1, hucep1, RRP-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23378 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O43159 | |
RRP8 | |
Between 406 and 456 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.