Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRP9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RRP9 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
RRP9 Polyclonal specifically detects RRP9 in Human samples. It is validated for Western Blot.Specifications
| RRP9 | |
| Polyclonal | |
| Purified | |
| RUO | |
| ribosomal RNA processing 9, small subunit (SSU) processome component, homolog(yeast), RNA, U3 small nucleolar interacting protein 2, RNU3IP2, RRP9 homolog, U3 small nucleolar ribonucleoprotein-associated 55 kDa protein, U3 small nucleolar RNA-interacting protein 2, U3 snoRNP-associated 55 kDa protein, U3 snoRNP-associated 55-kDa protein, U355K, U3-55KRRP9, small subunit (SSU) processome component, homolog | |
| RRP9 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| O43818 | |
| 9136 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title