Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558545 Supplier Invitrogen™ Supplier No. PA558545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THQLYSTSHD RSVKVWNVAE NSYVETLFGH QDAVAALDAL SRECCVTAGG RDGTVRVWKI PEESQLVFYG HQGSIDCIHL INEEHMVSGA DDGSVALWGL SKKRPL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The Ku autoantigen is a heterodimer of 70kDa (p70) and approximately 80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RRP9
Gene Accession No. O43818
Gene Alias 55kDa; D19435; D9Wsu10e; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast); ribosomal RNA processing 9, U3 small nucleolar RNA binding protein; RNA, U3 small nucleolar interacting protein 2; RNU3IP2; Rrp9; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; RRP9, small subunit (SSU) processome component, homolog (yeast); U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein; U3 snoRNP-associated protein; U355K; U3-55k
Gene Symbols RRP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RRP9. Recombinant protein control fragment (Product #RP-92413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.