Learn More
Invitrogen™ RRP9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110330
Description
Immunogen sequence: PDDSAIFSAA KDCSIIKWSV ESGRKLHVIP RAKKGAEGKP PGHSSHVLCM AISSDGKYLA SGDRSKLILI WEAQSCQHLY TFTGHRDAVS GLAFRRGTHQ LYSTSHDRSV KVWNVAENSY VETLFGHQDA VAALDALSRE CCVTAGGRDG TVRVWKIPEE SQLVFYGHQG SIDCIHLINE EHMVSGADDG SVALWGLSKK RPLALQREAH GLRGEPGLEQ PFWISSVAAL LNTDLVATGS HSSCVRLWQC GEGFRQLDLL CDIPLVGFIN SLKFSSSGDF LVAGVGQEHR LGRWWRIKEA RNSVCIIPLR RVPVPPAAGS.
The Ku autoantigen is a heterodimer of 70kDa (p70) and approximately 80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.
Specifications
RRP9 | |
Polyclonal | |
Unconjugated | |
RRP9 | |
55kDa; D19435; D9Wsu10e; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog; ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast); ribosomal RNA processing 9, U3 small nucleolar RNA binding protein; RNA, U3 small nucleolar interacting protein 2; RNU3IP2; Rrp9; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; RRP9, small subunit (SSU) processome component, homolog (yeast); U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein; U3 snoRNP-associated protein; U355K; U3-55k | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27966 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q91WM3 | |
RRP9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 156-475 of human RRP9 (NP_004695.1). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.