Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRP9 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566558
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6558 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6558 Supplier Bethyl Laboratories, Inc Supplier No. A303970AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

RRP9 contains 7 WD repeats and belongs to the WD repeat RRP9 family. It is a component of a nucleolar small nuclear ribonucleoprotein particle (snoRNP) thought to participate in the processing and modification of pre-ribosomal RNA.

Specifications

Antigen RRP9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RRP9
Gene Accession No. O43818
Gene Alias RNA, U3 small nucleolar interacting protein 2, RNU3IP2, RRP9 homolog, RRP9, small subunit (SSU) processome component, homolog, U3 small nucleolar ribonucleoprotein-associated 55 kDa protein, U3 snoRNP-associated 55 kDa protein, U3 snoRNP-associated 55-kDa protein, U3-55K
Gene Symbols RRP9
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9136
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.