Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563144 Supplier Invitrogen™ Supplier No. PA563144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHPTGHQSKE ELGRAMQVAK VSTASVGRFQ ERLPKEKVPR GSGKKRKFQP LFGDFAAEKK NQLELLRVMN SKKP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rrs1
Gene Accession No. Q15050
Gene Alias 5730466A07Rik; AA415037; D1Ertd701e; homolog of yeast ribosome biogenesis regulator; homolog of yeast ribosome biogenesis regulatory protein RRS1; KIAA0112; MNCb-2643; regulator for ribosome resistance homolog; ribosome biogenesis regulator homolog; ribosome biogenesis regulatory protein homolog; ribosome biogenesis regulatory protein RRS1 homolog; Rrr; Rrs1; RRS1 ribosome biogenesis regulator homolog; RRS1 ribosome biogenesis regulator homolog (S. cerevisiae)
Gene Symbols Rrs1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RRS1. Recombinant protein control fragment (Product #RP-93929).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.