Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRUT-3 Monoclonal Antibody (ZY203), Abbomax

Catalog No. 502204427
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4427 Supplier Abbomax Inc Supplier No. 606230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Colon tissue

RRUT-3 Monoclonal antibody specifically detects RRUT-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RRUT-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ZY203
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TIMP1
Gene Accession No. P01033
Gene Alias CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1
Host Species Mouse
Immunogen A recombinant protein TIMP-1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.