Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RS 246204
SDP

Catalog No. 885350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-535-0 50 mg
88-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-535-0 Supplier Tocris Bioscience™ Supplier No. 8853/50
Add to Cart
Add to Cart

R-Spondin-1 substitute used to initiate small intestinal organoids

RS 246204 is a R-Spondin-1 substitute. Able to initiate formation and growth of mouse intestinal organoids in media without R-Spondin-1 protein. In vivo, RS 24620 has regenerative effect in damaged intestinal epithelium mouse model.

Specifications

Chemical Name or Material 3,7-Dihydro-3,7-dimethyl-8-(2-quinolinylthio)-1H-purine-2,6-dione
CAS 878451-87-7
Quantity 50 mg
Target Compounds for Stem Cell Proliferation & Renewal
Molecular Formula C16H13N5O2S
Product Type Bioactive Small Molecule
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.