Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(RS)-3,4-DCPG, Tocris Bioscience™

Catalog No. p-4783107
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent systemically active anticonvulsant. Racemate of (R)-3,4,-DCPG (Cat. No. 1395)

Systemically active anticonvulsant that is 30-100-fold more potent in vivo than the separate enantiomers (S)-3,4-DCPG or (R)-3,4,-DCPG.

S-enantiomer and R-enantiomer also available.

Chemical Identifiers

CAS 176796-64-8
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
InChI Key IJVMOGKBEVRBPP-UHFFFAOYSA-N
Synonym rs-3,4-dcpg, rs-3,4-dicarboxyphenylglycine, dcpg, 4-1-amino-2-hydroxy-2-oxoethyl phthalic acid, 1,2-benzenedicarboxylic acid, 4-aminocarboxymethyl
PubChem CID 3313698
IUPAC Name 4-[amino(carboxy)methyl]phthalic acid
SMILES C1=CC(=C(C=C1C(C(=O)O)N)C(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 239.18
Percent Purity >99%
Chemical Name or Material (RS)-3, 4-DCPG
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.