Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (RS)-(±)-Sulpiride
GREENER_CHOICESDP

Catalog No. 0894100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-941-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-941-00 Supplier R&D Systems™ Supplier No. 0894/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Standard selective D2-like antagonist

Standard D2-like dopamine receptor antagonist.

S-enantiomer also available.

Chemical Identifiers

CAS 15676-16-1
Molecular Formula C15H23N3O4S

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 341.42
Purity >99%
Molecular Weight (g/mol) 341.43
Chemical Name or Material (RS)-(±)-Sulpiride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.