Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (RS)-(Tetrazol-5-yl)glycine
GREENER_CHOICE

Catalog No. 031210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-121-0 10 mg
03-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 03-121-0 Supplier Tocris Bioscience™ Supplier No. 0312/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent NMDA agonist

(RS)-(Tetrazol-5-yl)glycine is a potent NMDA receptor agonist, approximately 20 times more active than NMDA.

Specifications

CAS 138199-51-6
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 50 mM in water
Target NMDA Receptor Agonists
Molecular Formula C3H5N5O2
Formula Weight Observed MW: 143.1
Purity 98%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.