Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius RS232 Interface Data Cable
GREENER_CHOICE

Catalog No. 14558849 Shop All Sartorius Products
Change view
Click to view available options
Connection Type:
RS232 9-pin (male) to RS232 9-pin (female)
RS232 9-pin (male) to USB type A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
14-558-849 RS232 9-pin (male) to USB type A
14-558-851 RS232 9-pin (male) to RS232 9-pin (female)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-849 Supplier Sartorius Supplier No. YCCD09MUSBA
Add to Cart
Add to Cart

RS232 Interface Data Cable for Entris™ Lab Balance.

  • To connect Sartorius laboratory balances of the product family Entris™ to an external device via RS232 interface
  • Color: Black
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Connection of a Secura, Quintix or Practum to computer, printer or second display.
Connection Type RS232 9-pin (male) to USB type A
Length (English) 5 ft.
Length (Metric) 1.5 m
Type Data Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.