Learn More
Invitrogen™ RSAD1 Polyclonal Antibody

Description
Immunogen sequence: VTLEANPTSA PGSRLAEFGA AGVNRLSIGL QSLDDTELRL LGRTHSACDA LRTLAEARRL FPGRVSVDLM LGLPAQQVGP WLGQLQELLH HC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 27%.
Specifications
Specifications
| Antigen | RSAD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RSAD1 |
| Gene Accession No. | Q9HA92 |
| Gene Alias | B430319G23; BC056485; oxygen-independent coproporphyrinogen-III oxidase-like protein RSAD1; Putative heme chaperone; radical S-adenosyl methionine domain containing 1; radical S-adenosyl methionine domain-containing protein 1, mitochondrial; RSAD1 |
| Gene Symbols | RSAD1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.