Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSBN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561986 Supplier Invitrogen™ Supplier No. PA561986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSTSALFTFS PLTVSAAGPK HKGHKERHKH HHHRGPDGDP SSCGTDLKHK DKQENGERTG GVPLIKA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Histone demethylase that specifically demethylates dimethylated 'Lys-20' of histone H4 (H4K20me2), thereby modulating chromosome architecture. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSBN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSBN1
Gene Accession No. Q5VWQ0
Gene Alias C230004D03Rik; E330004N01; KDM9; Kiaa3002; Lysine-specific demethylase 9; mKIAA3002; RGD1307231; Rosbin; rosbin, round spermatid basic protein 1; round spermatid basic protein 1; Rsbn; Rsbn1; Rsbp
Gene Symbols RSBN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSBN1. Recombinant protein control fragment (Product #RP-104024).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.