Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566088 Supplier Invitrogen™ Supplier No. PA566088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDLETLKEDS EFTKVEMDNL DNAQTSGIEE PSETKGSMQK SKFKYKLVPE EETTASENTE ITSERQKEGI KLTIRI.

RSF1 is required for assembly of regular nucleosome arrays by the RSF chromatin remodelling complex. Rsf1 facilitates transcription of hepatitis B virus (HBV) genes by the pX transcription activator. In case of infection by HBV, together with pX, it represses TNF-alpha induced NF-kappaB transcription activation. Rsf1 represses transcription when artificially recruited to chromatin by fusion to a heterogeneous DNA binding domain. Rsf1 interacts with SMARCA5/SNF2H to form the RSF complex and also binds the HBV pX/HBx protein, which is required to activate transcription of the viral genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSF1
Gene Accession No. Q96T23
Gene Alias 4832420A03Rik; C030033M12Rik; Gm164; HBV pX associated protein-8; HBV pX-associated protein 8; HBXAP; hepatitis B virus x associated protein; hepatitis B virus x-associated protein; p325; p325 subunit of RSF chromatin-remodeling complex; Remodeling and spacing factor 1; RSF1; RSF-1; XAP8
Gene Symbols RSF1
Host Species Rabbit
Immunogen Recombinant Human RSF1. Recombinant protein control fragment (Product #RP-105074).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.