Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570802 Supplier Invitrogen™ Supplier No. PA570802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Yeast: 83%; Zebrafish: 83%.

Potential effector of the planar cell polarity signaling pathway. Plays a role in targeted membrane trafficking most probably at the level of vesicle fusion with membranes. Involved in cilium biogenesis by regulating the transport of cargo proteins to the basal body and to the apical tips of cilia. More generally involved in exocytosis in secretory cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RSG1
Gene Accession No. A2A825
Gene Alias 6330545A04Rik; C1orf89; Ciliogenesis and planar polarity effector 2; ciliogenesis and planar polarity effector 2; REM2- and Rab-like small GTPase 1; CPLANE2; Gm723; im:7136213; miro domain-containing protein C1orf89; miro domain-containing protein C1orf89 homolog; Rem/Rab-Similar GTPase 1; REM2 and RAB like small GTPase 1; REM2- and Rab-like small GTPase 1; REM2 and RAB-like small GTPase 1; RGD1561421; RSG1; zgc:101035; zgc:101035 protein; zgc:113864
Gene Symbols RSG1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse RSG1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76166
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.