Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555861
Description
Immunogen sequence: KFDQYMHTDV PERDLTAFRQ AWELPLLRVK SVPGRRLADG RTLDGRAGLA DVAHILNGLA EQLWHQDQVA AGLLPNPPES APE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Potential effector of the planar cell polarity signaling pathway. Plays a role in targeted membrane trafficking most probably at the level of vesicle fusion with membranes. Involved in cilium biogenesis by regulating the transport of cargo proteins to the basal body and to the apical tips of cilia. More generally involved in exocytosis in secretory cells.
Specifications
RSG1 | |
Polyclonal | |
Unconjugated | |
RSG1 | |
6330545A04Rik; C1orf89; Ciliogenesis and planar polarity effector 2; ciliogenesis and planar polarity effector 2; REM2- and Rab-like small GTPase 1; CPLANE2; Gm723; im:7136213; miro domain-containing protein C1orf89; miro domain-containing protein C1orf89 homolog; Rem/Rab-Similar GTPase 1; REM2 and RAB like small GTPase 1; REM2- and Rab-like small GTPase 1; REM2 and RAB-like small GTPase 1; RGD1561421; RSG1; zgc:101035; zgc:101035 protein; zgc:113864 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79363 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BU20 | |
RSG1 | |
Recombinant protein corresponding to Human RSG1. Recombinant protein control fragment (Product #RP-94206). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction