Learn More
Promega RSK1 Kinase Enzyme System, 10microg
Catalog No. PRV4046 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4046 10 μg
1 options
Catalog No. PRV4046 Supplier Promega Supplier No. V4046
Recombinant full-length human RSK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RSK1 is a member of the RSK (ribosomal S6 kinase) family that consists of growth factor-regulated serine/threonine kinases.
  • Easily Screen and Profile RSK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C