Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSK1, p Ser380 Antibody (1024F) - Azide and BSA Free, R&D Systems™

Rabbit Monoclonal Antibody
$243.50 - $487.50
Specifications
| Antigen | RSK1 (p Ser380) |
|---|---|
| Clone | 1024F |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.25 ug/ml, Immunocytochemistry 0.3 - 1 ug/ml |
| Applications | Western Blot, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB79672SP
![]() |
Novus Biologicals
MAB79672SP |
25 μg |
Each for $243.50
|
|
|||||
MAB79672
![]() |
Novus Biologicals
MAB79672 |
100 μg |
Each for $487.50
|
|
|||||
Description
RSK1 Monoclonal specifically detects RSK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| RSK1 (p Ser380) | |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS with No Preservative | |
| 6195 | |
| Phosphopeptide containing the human RSK1 S380 site Accession # Q15418 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1024F | |
| Western Blot 0.25 ug/ml, Immunocytochemistry 0.3 - 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1 | |
| RPS6KA1 | |
| IgG | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title