Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSK1, p Ser380 Antibody (1E11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | RSK1 |
---|---|
Clone | 1E11 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
RSK1 Monoclonal antibody specifically detects RSK1 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
RSK1 | |
Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1 | |
A synthesized peptide derived from Human RSK1 [p Ser380] [UniProt Q15418] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1E11 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6195 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title