Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSK1/RSK2/RSK3 Monoclonal Antibody (F.940.7)
GREENER_CHOICE

Catalog No. PIMA515040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515040 Supplier Invitrogen™ Supplier No. MA515040
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK1/RSK2/RSK3
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F.940.7
Concentration 67 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Rps6ka1
Gene Accession No. P18653, P18654, P51812, Q15349, Q15418, Q63531, Q9WUT3
Gene Alias 90 kDa ribosomal protein S6 kinase 1; 90 kDa ribosomal protein S6 kinase 2; 90 kDa ribosomal protein S6 kinase 3; 90kDa; C130006E23; CLS; D17Wsu134e; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); HU-1; HU-2; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPK-activated protein kinase 1b; MAPK-activated protein kinase 1c; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAP kinase 1b; MAPKAP kinase 1C; MAPKAPK-1; MAPKAPK1A; MAPKAPK-1a; MAPKAPK1B; MAPKAPK-1b; Mapkapk1c; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; MPK-9; MRX19; p90 rsk; p90Rsk; p90-RSK; p90-RSK 1; p90-RSK 2; p90-RSK 3; p90Rsk1; p90-Rsk1; p90RSK2; p90-RSK2; p90RSK3; p90-RSK3; p90S6K; pp90rsk; pp90RSK1; pp90RSK2; pp90RSK3; protein-tyrosine kinase Mpk-9; RGD1563860; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase alpha 1; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase alpha-3; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal protein S6 kinase, polypeptide 2; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; Ribosomal S6 kinase 2; ribosomal S6 kinase 3; RP11-393H10.3; RPS6KA; Rps6ka1; RPS6KA1L; Rps6ka2; Rps6ka3; Rps6ka-rs1; RSK; Rsk1; Rsk-1; RSK2; RSK-2; RSK3; RSK-3; S6 protein kinase (Rsk-1); S6K-alpha; S6K-alpha 1; S6K-alpha 2; S6K-alpha-1; S6K-alpha2; S6K-alpha-2; S6K-alpha3; S6K-alpha-3; S6KII-alpha
Gene Symbols Rps6ka1, RPS6KA2, RPS6KA3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Asp659 of human p90RSK.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100721226, 100728619, 100729439, 110651, 117269, 20111, 20112, 501560, 6195, 6196, 6197, 81771
Target Species Human, Mouse, Rat, Monkey, Guinea Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.