Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSK2 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130107FR
Description
RSK2 Polyclonal antibody specifically detects RSK2 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
RSK2 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
90 kDa ribosomal protein S6 kinase 3, CLS, Coffin-Lowry syndrome, EC 2.7.11, EC 2.7.11.1, HU-3, Insulin-stimulated protein kinase 1, ISPK1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, MAPKAPK1B, mental retardation, X-linked 19, MRX19, p90-RSK2, p90RSK3, pp90RSK2, ribosomal protein S6 kinase alpha-3, ribosomal protein S6 kinase, 90kD, polypeptide 3, ribosomal protein S6 kinase, 90kDa, polypeptide 3, Ribosomal S6 kinase 2, RSK, RSK-2, RSK2p90-RSK 3, S6K-alpha3, S6K-alpha-3 | |
maps to a region between residue 1 and 50 of human ribosomal S6 kinase 2 using the numbering given in entry NP_004577.1 | |
0.1 mL | |
Cancer, Phospho Specific, Protein Kinase | |
6197 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction