Learn More
RSK2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00683

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine, Dog, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Chimpanzee and Naked
RSK3 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).Specifications
RSK2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P51812 | |
RPS6KA3 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RPS6KA3 | |
Ribosomal protein S6 kinase alpha-3; S6K-alpha 3; 90 kDa ribosomal protein S6 kinase 3; p90-RSK 3; Ribosomal S6 kinase 2; RSK-2; pp90RSK2; Insulin-stimulated protein kinase 1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPKAPK1B; RPS6KA3; ISPK1; RSK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6197 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.