Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSK3 Monoclonal Antibody (3C4C8)
GREENER_CHOICE

Catalog No. PIMA538625
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538625 Supplier Invitrogen™ Supplier No. MA538625
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

RPS6KA2 (RSK3) serine/threonine kinase is a member of the ribosomal S6 kinase (RSK) family. It is believed that RPS6KA2 is a signal-transducing intermediate in the cellular response to several growth factors. It is found in many tissues, but in higher levels in lung and skeletal muscle. The p90 ribosomal S6 kinases (RSK)1-4 are downstream members of the extracellular signal-regulated kinase (ERK)/MAPK cascade. The loss of RSK2 activity in humans leads to Coffin-Lowry syndrome, which is characterized by mental retardation and growth deficits and may have a tonic regulation on G-protein coupled signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3C4C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RPS6KA2
Gene Accession No. Q15349, Q9WUT3
Gene Alias 90 kDa ribosomal protein S6 kinase 2; 90kDa; D17Wsu134e; HU-2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90rsk; p90-RSK 2; p90RSK2; p90-RSK3; pp90rsk; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; ribosomal S6 kinase 3; Rps6ka2; Rps6ka-rs1; RSK; Rsk3; RSK-3; S6K-alpha; S6K-alpha 2; S6K-alpha2; S6K-alpha-2
Gene Symbols RPS6KA2
Host Species Mouse
Immunogen Purified recombinant fragment of human RPS6KA2 (aa: 415-734) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20112, 6196
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.