Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536328 Supplier Invitrogen™ Supplier No. PA536328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 84 kDa. Purity is >95% by SDS-PAGE.

RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains. RSK4 encodes a member of ribosomal S6 kinase family, serine-threonine protein kinases which are regulated by growth factors. The encoded protein may be distinct from other members of this family, however, as studies suggest it is not growth factor dependent and may not participate in the same signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Rps6ka6
Gene Accession No. Q7TPS0, Q9UK32
Gene Alias 2610524K04Rik; 90 kDa ribosomal protein S6 kinase 6; p90-RSK 6; p90RSK6; pp90RSK4; RGD1560817; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal S6 kinase 4; Rps6ka6; RSK4; RSK-4; S6KA6; S6K-alpha 6; S6K-alpha-6
Gene Symbols Rps6ka6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 653-700 of human Rsk-4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27330, 317203, 67071
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.