Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590097
Description
Immunogen sequence: GGNWDNISDG AKDLLSHMLH MDPHQRYTAE QILKHSWITH RDQLPNDQPK RNDVSHVVKG AMVATYSALT HKTFQPVLEP VAASSLAQRR SMKKRTSTGL; Positive Samples: Mouse kidney; Cellular Location: Cytoplasm, Nucleus, cytosol.
RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains. RSK4 encodes a member of ribosomal S6 kinase family, serine-threonine protein kinases which are regulated by growth factors. The encoded protein may be distinct from other members of this family, however, as studies suggest it is not growth factor dependent and may not participate in the same signaling pathways.
Specifications
RSK4 | |
Polyclonal | |
Unconjugated | |
Rps6ka6 | |
2610524K04Rik; 90 kDa ribosomal protein S6 kinase 6; p90-RSK 6; p90RSK6; pp90RSK4; RGD1560817; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal S6 kinase 4; Rps6ka6; RSK4; RSK-4; S6KA6; S6K-alpha 6; S6K-alpha-6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67071 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.47 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7TPS0 | |
Rps6ka6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 646-745 of human RPS6KA6 (NP_0553111). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction