Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSL1D1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151999
Description
RSL1D1 Polyclonal specifically detects RSL1D1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
RSL1D1 | |
Polyclonal | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_056474.2 | |
RSL1D1 | |
Peptide with sequence C-PTARKQLDKEQVRK corresponding to internal region according to NP_056474.2. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CATX11, CATX-11, Cellular senescence-inhibited gene protein, CSIGMGC142259, DKFZP564M182, L12, MGC138433, PBK1cellular senescence inhibited gene protein, Protein PBK1, ribosomal L1 domain containing 1, ribosomal L1 domain-containing protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
26156 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction