Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSL1D1 Polyclonal Antibody, Bioss
SDP

Catalog No. 502280061
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0061 Supplier Bioss Supplier No. BS22952R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSL1D1 Polyclonal antibody specifically detects RSL1D1 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RSL1D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene RSL1D1
Gene Accession No. Q8BVY0
Gene Alias 2410005K20Rik; Ac1158; C76439; CATX11; CATX-11; cellular senescence inhibited; cellular senescence inhibited gene protein; cellular senescence-inhibited gene protein; CSIG; DKFZP564M182; L12; MGC138433; MGC142259; pBK1; Protein PBK1; ribosomal L1 domain containing 1; ribosomal L1 domain-containing protein 1; RSL1D1; UTP30
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse RSL1D1
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302898, 66409
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.