Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSL1D1 Polyclonal Antibody, HRP, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS22952RHRP

This item is not returnable.
View return policy
Description
RSL1D1 Polyclonal antibody specifically detects RSL1D1 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
RSL1D1 | |
Polyclonal | |
HRP | |
RSL1D1 | |
2410005K20Rik; Ac1158; C76439; CATX11; CATX-11; cellular senescence inhibited; cellular senescence inhibited gene protein; cellular senescence-inhibited gene protein; CSIG; DKFZP564M182; L12; MGC138433; MGC142259; pBK1; Protein PBK1; ribosomal L1 domain containing 1; ribosomal L1 domain-containing protein 1; RSL1D1; UTP30 | |
KLH conjugated synthetic peptide derived from mouse RSL1D1 | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q8BVY0 | |
Rabbit | |
Protein A | |
RUO | |
302898, 66409 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction