Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSL24D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564114 Supplier Invitrogen™ Supplier No. PA564114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCYFCSGPIY PGHGMMFVRN DCKVFRFCKS KCHKNFKKKR NPRKVRWTKA FRKAAGKELT VDNSFEFEKR RNEPIKYQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a protein sharing a low level of sequence similarity with human ribosomal protein L24. Although this gene has been referred to as RPL24, L30, and 60S ribosomal protein L30 isolog in the sequence databases, it is distinct from the human genes officially named RPL24 (which itself has been referred to as ribosomal protein L30) and RPL30. The protein encoded by this gene localizes to the nucleolus and is thought to play a role in the biogenesis of the 60S ribosomal subunit. The precise function of this gene is currently unknown. This gene utilizes alternative polyadenylation signals and has multiple pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSL24D1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSL24D1
Gene Accession No. Q9UHA3
Gene Alias 2410159K22Rik; 60S ribosomal protein L30 isolog; C15orf15; homolog of yeast ribosomal like protein 24; HRP-L30-iso; L30; My024; my024 protein; probable ribosome biogenesis protein RLP24; ribosomal L24 domain containing 1; ribosomal L24 domain-containing protein 1; Ribosomal protein L24-like; RLP24; RPL24; RPL24L; RSL24D1; TVAS3
Gene Symbols RSL24D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSL24D1. Recombinant protein control fragment (Product #RP-103509).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.