Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPH10B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570183 Supplier Invitrogen™ Supplier No. PA570183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Rat: 91%.

RSPH10B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH10B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RSPH10B
Gene Accession No. P0C881
Gene Alias 4930526H21Rik; hypothetical protein LOC222967; LOC100725924; Radial spoke head 10 homolog B; radial spoke head 10 homolog B (Chlamydomonas); radial spoke head 10 homolog B; radial spoke head 10 homolog B2; radial spoke head 10 homolog B2; RGD1311893; Rsph10b; RSPH10B2
Gene Symbols RSPH10B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RSPH10B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222967
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.