Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPH10B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116667 Supplier Invitrogen™ Supplier No. PA5116667
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Jurkat Immunogen sequence: MVKEKKKADK KGEKSARSPS SLSDNLDFSK QDGNTTRQEM SPAGVPLLGM QLNEVKPKKD RQNVQQNEDA TQYEESILTK LIVESYEGEK VRGLYEGEGF AAFQGGCTYR GMFSEGLMHG QGTYIWADGL KYEGDFVKNV PMNHGVYTWP DGSMYEGEVV NGMRNGFGMF KCSTQPVSYI GHWCNGKRHG KGSIYYNQEG TCWYEGDWVQ NIKKGWGIRC YKSGNIYEGQ WEDNMRHGEG RMRWLTTNEE YTGRWERGIQ NGFGTHTWFL KRIRSSQYPL RNEYIGEFVN GYRHGRGKFY.

RSPH10B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH10B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RSPH10B
Gene Accession No. P0C881
Gene Alias 4930526H21Rik; hypothetical protein LOC222967; LOC100725924; Radial spoke head 10 homolog B; radial spoke head 10 homolog B (Chlamydomonas); radial spoke head 10 homolog B; radial spoke head 10 homolog B2; radial spoke head 10 homolog B2; RGD1311893; Rsph10b; RSPH10B2
Gene Symbols RSPH10B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RSPH10B (NP_7758363).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222967
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.