Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSPH4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556667 Supplier Thermo Scientific Supplier No. PA556667
Add to Cart
Add to Cart

RSPH4A Polyclonal Antibody

RSPH4A Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH4A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSPH4A
Gene Accession No. Q5TD94
Gene Alias A230081C05; CILD11; dJ412I7.1; radial spoke head 4 homolog A; radial spoke head 4 homolog A (Chlamydomonas); radial spoke head protein 4 homolog A; Radial spoke head-like protein 3; radial spokehead-like 3; RSHL3; Rsph4a; RSPH6B
Gene Symbols RSPH4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSPH4A. Recombinant protein control fragment (Product #RP-94637).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.