Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSPH4A/RSHL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981394
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1394 Supplier Bioss Supplier No. BS11472R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that appears to be a component the radial spoke head, as determined by homology to similar proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates. Radial spokes, which are regularly spaced along cilia, sperm, and flagella axonemes, consist of a thin ′stalk′ and a bulbous ′head′ that form a signal transduction scaffold between the central pair of microtubules and dynein. Mutations in this gene cause primary ciliary dyskinesia 1, a disease arising from dysmotility of motile cilia and sperm. Alternative splicing results in multiple transcript variants.

Specifications

Antigen RSPH4A/RSHL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RSPH4A
Gene Accession No. Q5TD94
Gene Alias CILD11; dJ412I7.1; radial spoke head 4 homolog A; Radial spoke head protein 4 homolog A; Radial spoke head-like protein 3; radial spokehead-like 3; RSHL3; RSPH4A; RSPH6B
Gene Symbols RSPH4A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RSPH4A/RSHL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345895
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.