Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPH6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571255 Supplier Invitrogen™ Supplier No. PA571255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rat: 93%.

The protein encoded by this gene is similar to a sea urchin radial spoke head protein. Radial spoke protein complexes form part of the axoneme of eukaryotic flagella and are located between the axoneme's outer ring of doublet microtubules and central pair of microtubules. In Chlamydomonas, radial spoke proteins are thought to regulate the activity of dynein and the symmetry of flagellar bending patterns. This gene maps to a region of chromosome 19 that is linked to primary ciliary dyskinesia-2 (CILD2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH6A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RSPH6A
Gene Accession No. Q9H0K4
Gene Alias Dmwd; dystrophia myotonica-containing WD repeat motif; hematological and neurological expressed 1 (Hn1-ps1) pseudogene; Hn1-ps1; ortholog of mouse radial spokehead-like 1; radial spoke head 6 homolog A; radial spoke head 6 homolog A (Chlamydomonas); radial spoke head protein 6 homolog A; Radial spoke head-like protein 1; radial spokehead-like 1; Rshl1; RSP4; RSP6; RSPH4B; RSPH6A
Gene Symbols RSPH6A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human RSPH6A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.