Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPH9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556821 Supplier Invitrogen™ Supplier No. PA556821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSVVKGRFMG DPSYEYEHTE LQKVNEGEKV FEEEIVVQIK EETRLVSVID QIDKAVAIIP RGALFKTPFG PTHVNRTFEG LSLSEAKKLS SY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

This gene encodes a protein thought to be a component of the radial spoke head in motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSPH9
Gene Accession No. Q9H1X1
Gene Alias 1700027N10Rik; C23H6orf206; C6orf206; CILD12; MRPS18AL1; radial spoke head 9 homolog; radial spoke head 9 homolog (Chlamydomonas); radial spoke head component 9; radial spoke head protein 9 homolog; RGD1310693; RSPH9; similar to CG31803-PA
Gene Symbols RSPH9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSPH9. Recombinant protein control fragment (Product #RP-94950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.