Learn More
Invitrogen™ RSPO1 Polyclonal Antibody

Description
A suggested positive control is rat spleen tissue lysate. PA5-20749 can be used with blocking peptide PEP-0863.
Specifications
Specifications
| Antigen | RSPO1 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RSPO1 |
| Gene Accession No. | Q2MKA7, Q9Z132 |
| Gene Alias | CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin |
| Gene Symbols | RSPO1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.