Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117129 Supplier Invitrogen™ Supplier No. PA5117129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, NCI-H460, HeLa, Mouse liver, Mouse skin, Mouse brain, Mouse spinal cord, Rat brain Immunogen sequence: MHLRLISWLF IILNFMEYIG SQNASRGRRQ RRMHPNVSQG CQGGCATCSD YNGCLSCKPR LFFALERIGM KQIGVCLSSC PSGYYGTRYP DINKCTKCKA.

RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPO3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RSPO3
Gene Accession No. Q2TJ95, Q9BXY4
Gene Alias 2810459H04Rik; AW742308; Cabriolet; cristin 1; Cristin1; cristin-1; cysteine-rich and single thrombospondin domain-containing protein 1; hPWTSR; hRspo3; Nucleopondin; protein with TSP type-1 repeat; PWTSR; RGD1563246; Roof plate-specific spondin-3; Rspo3; R-spondin 3; R-spondin 3 homolog; R-spondin 3 homolog (Xenopus laevis); R-spondin 3-like protein; R-spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain 2; thrombospondin, type I, domain containing 2; Thsd2; Unknown (protein for MGC:137973)
Gene Symbols RSPO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RSPO3 (NP_1161732).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498997, 72780, 84870
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.