Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSPO3 Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502280938
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0938 Supplier Bioss Supplier No. BS18877RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSPO3 Polyclonal antibody specifically detects RSPO3 in Human samples. It is validated for Western Blot

Specifications

Antigen RSPO3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene RSPO3
Gene Accession No. Q9BXY4
Gene Alias 2810459H04Rik; AW742308; Cabriolet; cristin 1; Cristin1; cristin-1; cysteine-rich and single thrombospondin domain-containing protein 1; hPWTSR; hRspo3; Nucleopondin; protein with TSP type-1 repeat; PWTSR; RGD1563246; Roof plate-specific spondin-3; Rspo3; R-spondin 3; R-spondin 3 homolog; R-spondin 3 homolog (Xenopus laevis); R-spondin 3-like protein; R-spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain 2; thrombospondin, type I, domain containing 2; Thsd2; Unknown (protein for MGC:137973)
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RSPO3
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84870
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.