Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Abnova™ RSPO4 (Human) ELISA Kit
Shop All Abnova Corporation Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 Kit
                            
                        
                    Description
RSPO4 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human R-Spondin-4 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Description | R-spondin family, member 4 | 
| Cross Reactivity | Human | 
| Target Species | Human | 
| For Use With (Application) | Quantitative | 
| Gene ID (Entrez) | 343637 | 
| Quantity | 1 Kit | 
| Regulatory Status | RUO | 
| Storage Requirements | Store at 4°C for six months. For long term storage store at -20°C. Avoid repeated freezing and thawing. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction