Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSPO4 Monoclonal Antibody (435203)
GREENER_CHOICE

Catalog No. PIMA524160
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524160 Supplier Invitrogen™ Supplier No. MA524160
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In Western blots, 50-100% cross-reactivity with recombinant human (rh) R-Spondin 4 is observed and no cross-reactivity with recombinant mouse (rm) R-Spondin 1, rmR-Spondin 3, or rhR-Spondin 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPO4
Applications Western Blot
Classification Monoclonal
Clone 435203
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene RSPO4
Gene Accession No. Q8BJ73
Gene Alias A730099F22; A930029K19Rik; C20orf182; CRISTIN4; Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; hRspo4; mCristin-4; roof plate-specific spondin-4; RSPO4; R-spondin 4; R-spondin family, member 4; R-spondin-4; R-spondin-like protein
Gene Symbols RSPO4
Host Species Rat
Immunogen E. coli-derived recombinant mouse R-Spondin 4 Tyr21-Pro197.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228770
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.