Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSPO4 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547579
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
Specifications
RSPO4 | |
Polyclonal | |
Unconjugated | |
RSPO4 | |
A730099F22; A930029K19Rik; C20orf182; CRISTIN4; Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; hRspo4; mCristin-4; roof plate-specific spondin-4; RSPO4; R-spondin 4; R-spondin family, member 4; R-spondin-4; R-spondin-like protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
228770 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8BJ73 | |
RSPO4 | |
E. coli-derived recombinant mouse R-Spondin 4 Tyr21-Pro197. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction