Learn More
Invitrogen™ RSRC1 Polyclonal Antibody

Description
A suggested positive control is HepG2 cell lysate. PA5-20899 can be used with blocking peptide PEP-1013.
Specifications
Specifications
| Antigen | RSRC1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RSRC1 |
| Gene Accession No. | Q5PPJ2, Q96IZ7, Q9DBU6 |
| Gene Alias | 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53 |
| Gene Symbols | RSRC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.