Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560828 Supplier Invitrogen™ Supplier No. PA560828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGKDKELHNI KRGESGNIKA GLEHLPPAEQ AKARLQLVLE AAAKADEALK AKERNEEEAK RRKEEDQATL VEQVKRVKEI EAIESD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 81%.

RSRC1 is a gene that encodes a protein belonging to the serine and arginine rich-related protein family. This protein plays a crucial role in both constitutive and alternative mRNA splicing. Research suggests that this gene may be linked to schizophrenia, and a pseudogene of RSRC1 is present on chromosome 9. Alternative splicing of RSRC1 leads to the formation of multiple transcript variants that encode distinct isoforms. Additionally, RSRC1 has been associated with various disorders, including Autosomal Recessive Non-Syndromic Intellectual Disability and Intellectual Developmental Disorder, Autosomal Recessive 70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSRC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSRC1
Gene Accession No. Q96IZ7
Gene Alias 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53
Gene Symbols RSRC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSRC1. Recombinant protein control fragment (Product #RP-100236).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.