Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577169 Supplier Invitrogen™ Supplier No. PA577169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RSRC1 is a gene that encodes a protein belonging to the serine and arginine rich-related protein family. This protein plays a crucial role in both constitutive and alternative mRNA splicing. Research suggests that this gene may be linked to schizophrenia, and a pseudogene of RSRC1 is present on chromosome 9. Alternative splicing of RSRC1 leads to the formation of multiple transcript variants that encode distinct isoforms. Additionally, RSRC1 has been associated with various disorders, including Autosomal Recessive Non-Syndromic Intellectual Disability and Intellectual Developmental Disorder, Autosomal Recessive 70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSRC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RSRC1
Gene Accession No. Q5PPJ2, Q96IZ7, Q9DBU6
Gene Alias 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53
Gene Symbols RSRC1
Host Species Rabbit
Immunogen Recombinant full length Human RSRC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361956, 51319, 66880
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.