Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSRC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110332
Description
Immunogen sequence: EHLPPAEQAK ARLQLVLEAA AKADEALKAK ERNEEEAKRR KEEDQATLVE QVKRVKEIEA IESDSFVQQT FRSSKEVKKS VEPSEVKQAT STSGPASAVA DPPSTEKEID PTSIPTAIKY QDDNSLAHPN LFIEKADAEE KWFKRLIALR QERLMGSPVA.
RSRC1 is a gene that encodes a protein belonging to the serine and arginine rich-related protein family. This protein plays a crucial role in both constitutive and alternative mRNA splicing. Research suggests that this gene may be linked to schizophrenia, and a pseudogene of RSRC1 is present on chromosome 9. Alternative splicing of RSRC1 leads to the formation of multiple transcript variants that encode distinct isoforms. Additionally, RSRC1 has been associated with various disorders, including Autosomal Recessive Non-Syndromic Intellectual Disability and Intellectual Developmental Disorder, Autosomal Recessive 70.
Specifications
| RSRC1 | |
| Polyclonal | |
| Unconjugated | |
| RSRC1 | |
| 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361956, 51319, 66880 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PPJ2, Q96IZ7, Q9DBU6 | |
| RSRC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 117-276 of human RSRC1 (NP_001258763.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction