Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSRC1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | RSRC1 |
|---|---|
| Concentration | 0.67 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4130
![]()
|
Proteintech Group Inc
238261AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The arginine/serine-rich coiled-coil 1 protein (RSRC1) was initially identified as a non-snRNP splicing factor that has a role in both the constitutive and alternative splicing of pre-mRNA molecules. In cellular nuclear extracts immunodepleted of RSRC1, the second step of pre-mRNA splicing is blocked; recombit RSRC1 is able to restore splicing activity. RSRC1 has also been reported to be associated with schizophrenia as well as a unique marker of bone marrow-derived stem cells that also express the cdc34 marker.Specifications
| RSRC1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96IZ7 | |
| 51319 | |
| RSRC1 Fusion Protein Ag20604 | |
| Primary | |
| -20°C | |
| RSRC1 |
| 0.67 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BM 011, RSRC1, SRrp53 | |
| RSRC1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title